Assay Details
Gene Symbol: TNFSF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205168.1 | NP_001192097.1 | 3 - 4 | 281 | 71 | |
GenBank mRNA | FJ915118.1 | - | 3 - 4 | 281 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
