Assay Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205169.1 | NP_001192098.1 | 1 - 2 | 282 | 76 | |
GenBank mRNA | DN371994.1 | - | 1 - 2 | 282 | 76 | |
EU909456.1 | - | 1 - 2 | 257 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
