Assay Details
Gene Symbol: MIR222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049439.1 | 1 | 42 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | divergent protein kinase domain 2B | DIPK2B | CXHXorf36 | -389.4kb | Sense | Chr. X - 38996003 - 39039589 on NCBI Build CanFam3.1 |
Downstream | microRNA mir-221 | MIR221 | cfa-mir-221 | 94.8kb | Sense | Chr. X - 39523890 - 39523951 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
