Assay Details
Gene Symbol: RWDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286853.1 | NP_001273782.1 | 2 - 3 | 320 | 96 | |
GenBank mRNA | AJ388553.1 | - | 2 - 3 | 313 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
