Assay Details
Gene Symbol: ACYP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287168.1 | NP_001274097.1 | 2 - 3 | 148 | 125 | |
XM_005623670.3 | XP_005623727.1 | 2 - 3 | 223 | 125 | ||
GenBank mRNA | CO633742.1 | - | 1 - 2 | 434 | 125 | |
DT536013.1 | - | 2 - 3 | 148 | 125 | ||
DT540937.1 | - | 2 - 3 | 183 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001287168.1, XM_005623670.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
