Assay Details
Gene Symbol: PPP1R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313823.1 | NP_001300752.1 | 567 | 84 | ||
GenBank mRNA | DN415995.1 | - | 567 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
