Assay Details
Gene Symbol: eci2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002645.3 | NP_001002645.3 | 3 - 4 | 415 | 131 | |
XM_021470136.1 | XP_021325811.1 | 3 - 4 | 326 | 131 | ||
XM_021470137.1 | XP_021325812.1 | 3 - 4 | 405 | 131 | ||
GenBank mRNA | BC076531.1 | - | 3 - 4 | 342 | 131 | |
BC155309.1 | - | 6 - 7 | 561 | 129 | ||
EH435933.1 | - | 3 - 4 | 367 | 131 | ||
EH589679.1 | - | 3 - 4 | 361 | 131 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC155309.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
