Assay Details
Gene Symbol: lpar2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003578.2 | NP_001003578.1 | 2 - 3 | 1055 | 121 | |
XM_009299773.3 | XP_009298048.1 | 2 - 3 | 1053 | 121 | ||
GenBank mRNA | BC078264.1 | - | 2 - 3 | 1059 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
