Assay Details
Gene Symbol: ropn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004624.1 | NP_001004624.1 | 3 - 4 | 444 | 86 | |
GenBank mRNA | BC081402.1 | - | 3 - 4 | 444 | 86 | |
BC152162.1 | - | 3 - 4 | 447 | 86 | ||
BC165043.1 | - | 2 - 3 | 285 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
