Assay Details
Gene Symbol: pgam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007323.2 | NP_001007324.2 | 2 - 3 | 681 | 102 | |
XM_005165077.3 | XP_005165134.1 | 2 - 3 | 675 | 102 | ||
GenBank mRNA | BC085627.1 | - | 2 - 3 | 426 | 102 | |
BC095654.1 | - | 2 - 3 | 632 | 102 | ||
BC164655.1 | - | 2 - 3 | 406 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
