Assay Details
Gene Symbol: ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131309.3 | NP_571384.1 | 3 - 4 | 289 | 100 | |
XM_005173094.4 | XP_005173151.1 | 3 - 4 | 276 | 100 | ||
GenBank mRNA | BC050517.1 | - | 3 - 4 | 195 | 100 | |
BC058047.1 | - | 2 - 3 | 160 | 100 | ||
EH499718.1 | - | 3 - 4 | 253 | 100 | ||
U61395.1 | - | 2 - 3 | 166 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
