Assay Details
Gene Symbol: ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131309.3 | NP_571384.1 | 4 - 5 | 424 | 135 | |
XM_005173094.4 | XP_005173151.1 | 4 - 5 | 411 | 135 | ||
GenBank mRNA | BC050517.1 | - | 4 - 5 | 330 | 135 | |
BC058047.1 | - | 3 - 4 | 295 | 135 | ||
EH499718.1 | - | 4 - 5 | 388 | 135 | ||
U61395.1 | - | 3 - 4 | 301 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
