Assay Details
Gene Symbol: ptk2ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131796.1 | NP_571871.1 | 11 - 12 | 1180 | 86 | |
XM_005158162.4 | XP_005158219.1 | 10 - 11 | 86 | |||
XM_005158163.4 | XP_005158220.1 | 10 - 11 | 86 | |||
XM_005158164.4 | XP_005158221.1 | 10 - 11 | 86 | |||
XM_005158165.4 | XP_005158222.1 | 10 - 11 | 86 | |||
XM_009292411.3 | XP_009290686.1 | 10 - 11 | 86 | |||
XM_021466598.1 | XP_021322273.1 | 10 - 11 | 86 | |||
XM_021466599.1 | XP_021322274.1 | 10 - 11 | 86 | |||
XM_021466600.1 | XP_021322275.1 | 10 - 11 | 86 | |||
XM_021466601.1 | XP_021322276.1 | 10 - 11 | 86 | |||
XM_021466602.1 | XP_021322277.1 | 11 - 12 | 86 | |||
XM_021466603.1 | XP_021322278.1 | 10 - 11 | 86 | |||
XM_021466604.1 | XP_021322279.1 | 10 - 11 | 86 | |||
XM_021466605.1 | XP_021322280.1 | 10 - 11 | 86 | |||
XM_021466606.1 | XP_021322281.1 | 10 - 11 | 86 | |||
XM_021466607.1 | XP_021322282.1 | 10 - 11 | 86 | |||
XM_021466608.1 | XP_021322283.1 | 10 - 11 | 86 | |||
XM_021466609.1 | XP_021322284.1 | 11 - 12 | 86 | |||
GenBank mRNA | AF348085.1 | - | 11 - 12 | 1180 | 86 | |
BC055493.1 | - | 11 - 12 | 971 | 86 | ||
BC163886.1 | - | 9 - 10 | 993 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
