Assay Details
Gene Symbol: aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199690.2 | NP_955984.2 | 2 - 3 | 289 | 72 | |
XM_005173083.4 | XP_005173140.1 | 2 - 3 | 331 | 72 | ||
XM_009307693.3 | XP_009305968.1 | 2 - 3 | 334 | 72 | ||
GenBank mRNA | BC059643.1 | - | 2 - 3 | 287 | 72 | |
BC065447.1 | - | 2 - 3 | 289 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
