Assay Details
Gene Symbol: aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199690.2 | NP_955984.2 | 3 - 4 | 445 | 96 | |
XM_005173083.4 | XP_005173140.1 | 3 - 4 | 487 | 96 | ||
XM_009307693.3 | XP_009305968.1 | 3 - 4 | 490 | 96 | ||
GenBank mRNA | BC059643.1 | - | 3 - 4 | 443 | 96 | |
BC065447.1 | - | 3 - 4 | 445 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
