Assay Details
Gene Symbol: pelo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201136.1 | NP_957430.1 | 2 - 3 | 241 | 74 | |
GenBank mRNA | BC049484.1 | - | 2 - 3 | 241 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
