Assay Details
Gene Symbol: atg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009914.2 | NP_001009914.2 | 7 - 8 | 874 | 72 | |
NM_205618.2 | NP_991181.2 | 7 - 8 | 821 | 72 | ||
GenBank mRNA | AY398310.1 | - | 7 - 8 | 779 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
