Assay Details
Gene Symbol: tuba7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002230.1 | NP_001002230.1 | 5 - 6 | 1162 | 90 | |
XM_021479153.1 | XP_021334828.1 | 4 - 5 | 1571 | 90 | ||
GenBank mRNA | BC072721.1 | - | 5 - 6 | 1162 | 90 | |
BC165338.1 | - | 4 - 5 | 1088 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
