Assay Details
Gene Symbol: adprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003453.2 | NP_001003453.1 | 4 - 5 | 623 | 93 | |
GenBank mRNA | BC078399.1 | - | 4 - 5 | 469 | 93 | |
BC164170.1 | - | 3 - 4 | 361 | 93 | ||
EH441896.1 | - | 4 - 5 | 621 | 93 | ||
EH557169.1 | - | 4 - 5 | 664 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
