Assay Details
Gene Symbol: bnip3la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012242.1 | NP_001012242.1 | 3 - 4 | 398 | 121 | |
GenBank mRNA | AY424951.1 | - | 3 - 4 | 398 | 121 | |
BC163834.1 | - | 3 - 4 | 410 | 121 | ||
BC163857.1 | - | 3 - 4 | 410 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
