Assay Details
Gene Symbol: bag5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034183.1 | NP_001029355.1 | 4 - 5 | 1142 | 83 | |
XM_005156686.4 | XP_005156743.2 | 4 - 5 | 1027 | 83 | ||
XM_021480648.1 | XP_021336323.1 | 4 - 5 | 1205 | 83 | ||
GenBank mRNA | BC097176.1 | - | 4 - 5 | 1142 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
