Assay Details
Gene Symbol: myclb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045142.1 | NP_001038607.1 | 2 - 3 | 930 | 64 | |
GenBank mRNA | BC124204.1 | - | 2 - 3 | 931 | 64 | |
BC164499.1 | - | 1 - 2 | 600 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
