Assay Details
Gene Symbol: bag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201000.2 | NP_957294.2 | 1 - 2 | 267 | 63 | |
GenBank mRNA | BC053303.1 | - | 223 | 63 | ||
EH495617.1 | - | 1 - 2 | 293 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
