Assay Details
Gene Symbol: bag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201000.2 | NP_957294.2 | 2 - 3 | 376 | 74 | |
GenBank mRNA | BC053303.1 | - | 332 | 74 | ||
EH495617.1 | - | 2 - 3 | 402 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
