Assay Details
Gene Symbol: ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213339.1 | NP_998504.1 | 2 - 3 | 454 | 67 | |
XM_005159766.3 | XP_005159823.1 | 2 - 3 | 326 | 67 | ||
XM_005159767.3 | XP_005159824.1 | 2 - 3 | 313 | 67 | ||
XM_017352221.2 | XP_017207710.1 | 1 - 2 | 129 | 67 | ||
GenBank mRNA | BC053174.1 | - | 2 - 3 | 454 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
