Assay Details
Gene Symbol: rel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001841.3 | NP_001001841.2 | 5 - 6 | 497 | 84 | |
GenBank mRNA | AY163837.1 | - | 5 - 6 | 434 | 84 | |
BC076403.1 | - | 5 - 6 | 501 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
