Assay Details
Gene Symbol: rel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001841.3 | NP_001001841.2 | 9 - 10 | 1108 | 76 | |
GenBank mRNA | AY163837.1 | - | 9 - 10 | 1045 | 76 | |
BC076403.1 | - | 9 - 10 | 1112 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
