Assay Details
Gene Symbol: psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002543.2 | NP_001002543.2 | 4 - 5 | 487 | 62 | |
NM_001328552.2 | NP_001315481.1 | 5 - 6 | 464 | 62 | ||
GenBank mRNA | BC076265.1 | - | 4 - 5 | 443 | 62 | |
EH468130.1 | - | 4 - 5 | 564 | 62 | ||
EH587434.1 | - | 5 - 6 | 488 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
