Assay Details
Gene Symbol: mogat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006083.2 | NP_001006083.2 | 3 - 4 | 492 | 83 | |
NM_001326700.1 | NP_001313629.1 | 3 - 4 | 603 | 83 | ||
GenBank mRNA | BC083237.1 | - | 3 - 4 | 492 | 83 | |
BC165118.1 | - | 2 - 3 | 264 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
