Assay Details
Gene Symbol: acot9.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037399.1 | NP_001032476.1 | 3 - 4 | 465 | 61 | |
GenBank mRNA | BC108006.1 | - | 3 - 4 | 465 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
