Assay Details
Gene Symbol: enah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013521.2 | NP_001013539.2 | 6 - 7 | 1427 | 101 | |
NM_001045028.3 | NP_001038493.1 | 7 - 8 | 2164 | 101 | ||
XM_009294710.3 | XP_009292985.1 | 7 - 8 | 2219 | 101 | ||
XM_009294712.3 | XP_009292987.1 | 7 - 8 | 1988 | 101 | ||
XM_009294713.3 | XP_009292988.1 | 7 - 8 | 2171 | 101 | ||
XM_009294714.3 | XP_009292989.1 | 5 - 6 | 1802 | 101 | ||
XM_009294715.3 | XP_009292990.1 | 6 - 7 | 1483 | 101 | ||
XM_009294716.3 | XP_009292991.1 | 6 - 7 | 1482 | 101 | ||
XM_021468414.1 | XP_021324089.1 | 7 - 8 | 1589 | 101 | ||
XM_021468415.1 | XP_021324090.1 | 7 - 8 | 1588 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
