Assay Details
Gene Symbol: tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076600.1 | NP_001070068.1 | 3 - 4 | 461 | 63 | |
GenBank mRNA | BC124227.1 | - | 3 - 4 | 461 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
