Assay Details
Gene Symbol: rps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173232.1 | NP_775339.1 | 3 - 4 | 354 | 99 | |
XM_021481330.1 | XP_021337005.1 | 3 - 4 | 384 | 99 | ||
GenBank mRNA | AF506223.1 | - | 3 - 4 | 354 | 99 | |
BC059443.1 | - | 3 - 4 | 347 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
