Assay Details
Gene Symbol: ndrg3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199797.1 | NP_956091.1 | 11 - 12 | 872 | 81 | |
XM_005161986.2 | XP_005162043.1 | 11 - 12 | 966 | 81 | ||
XM_009296689.1 | XP_009294964.1 | 11 - 12 | 966 | 81 | ||
XM_009296690.2 | XP_009294965.1 | 9 - 10 | 774 | 81 | ||
XM_009296691.2 | XP_009294966.1 | 11 - 12 | 966 | 81 | ||
XM_017353522.1 | XP_017209011.1 | 11 - 12 | 966 | 81 | ||
GenBank mRNA | BC044139.1 | - | 11 - 12 | 872 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
