Assay Details
Gene Symbol: ndrg3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199797.1 | NP_956091.1 | 13 - 14 | 977 | 77 | |
XM_005161986.2 | XP_005162043.1 | 13 - 14 | 1071 | 77 | ||
XM_009296689.1 | XP_009294964.1 | 13 - 14 | 1071 | 77 | ||
XM_009296690.2 | XP_009294965.1 | 11 - 12 | 879 | 77 | ||
XM_009296691.2 | XP_009294966.1 | 13 - 14 | 1071 | 77 | ||
XM_017353522.1 | XP_017209011.1 | 13 - 14 | 1071 | 77 | ||
GenBank mRNA | BC044139.1 | - | 13 - 14 | 977 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
