Assay Details
Gene Symbol: rpl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200047.1 | NP_956341.1 | 4 - 5 | 501 | 83 | |
GenBank mRNA | BC059533.1 | - | 4 - 5 | 501 | 83 | |
BC071352.1 | - | 5 - 6 | 517 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
