Assay Details
Gene Symbol: tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320515.1 | NP_001307444.1 | 4 - 5 | 511 | 64 | |
NM_200908.2 | NP_957202.1 | 4 - 5 | 511 | 64 | ||
XM_017353659.2 | XP_017209148.1 | 4 - 5 | 518 | 64 | ||
GenBank mRNA | BC045486.1 | - | 480 | 64 | ||
BC159223.1 | - | 4 - 5 | 505 | 64 | ||
EH487910.1 | - | 4 - 5 | 519 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
