Assay Details
Gene Symbol: tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200908.2 | NP_957202.1 | 7 - 8 | 810 | 73 | |
GenBank mRNA | BC045486.1 | - | 779 | 73 | ||
EH487910.1 | - | 7 - 8 | 818 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
