Assay Details
Gene Symbol: tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200908.2 | NP_957202.1 | 8 - 9 | 961 | 95 | |
GenBank mRNA | BC045486.1 | - | 930 | 95 | ||
EH600327.1 | - | 3 - 4 | 544 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
