Assay Details
Gene Symbol: cela1.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003450.1 | NP_001003450.1 | 3 - 4 | 248 | 106 | |
GenBank mRNA | BC076046.1 | - | 3 - 4 | 248 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
