Assay Details
Gene Symbol: cela1.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003450.1 | NP_001003450.1 | 4 - 5 | 369 | 74 | |
GenBank mRNA | BC076046.1 | - | 4 - 5 | 369 | 74 |
Gene Symbol: LOC100334908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002666189.6 | XP_002666235.1 | 4 - 5 | 406 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
