Assay Details
Gene Symbol: cela1.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003450.1 | NP_001003450.1 | 6 - 7 | 660 | 78 | |
GenBank mRNA | BC076046.1 | - | 6 - 7 | 660 | 78 |
Gene Symbol: LOC100334908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002666189.6 | XP_002666235.1 | 6 - 7 | 697 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
