Assay Details
Gene Symbol: rad23aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003739.1 | NP_001003739.1 | 4 - 5 | 646 | 112 | |
GenBank mRNA | BC079526.1 | - | 4 - 5 | 646 | 112 | |
BC164761.1 | - | 4 - 5 | 505 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
