Assay Details
Gene Symbol: fuom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004530.1 | NP_001004530.1 | 1 - 2 | 144 | 76 | |
GenBank mRNA | BC081387.1 | - | 1 - 2 | 144 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
