Assay Details
Gene Symbol: irak1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020771.1 | NP_001018607.1 | 1 - 2 | 326 | 94 | |
GenBank mRNA | BC095828.1 | - | 1 - 2 | 326 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
