Assay Details
Gene Symbol: cela1.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025187.1 | NP_001020358.1 | 7 - 8 | 818 | 98 | |
GenBank mRNA | BC096885.1 | - | 7 - 8 | 818 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
