Assay Details
Gene Symbol: atg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037124.1 | NP_001032201.1 | 4 - 5 | 429 | 69 | |
GenBank mRNA | BC107646.1 | - | 4 - 5 | 429 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
