Assay Details
Gene Symbol: lpar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320133.1 | NP_001307062.1 | 3 - 4 | 1022 | 63 | |
XM_005171058.4 | XP_005171115.1 | 3 - 4 | 1004 | 63 | ||
XM_021475647.1 | XP_021331322.1 | 3 - 4 | 1004 | 63 | ||
XM_021475649.1 | XP_021331324.1 | 3 - 4 | 1004 | 63 | ||
XM_021475650.1 | XP_021331325.1 | 3 - 4 | 1004 | 63 | ||
GenBank mRNA | BC109431.1 | - | 3 - 4 | 911 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
