Assay Details
Gene Symbol: ogfod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320174.1 | NP_001307103.1 | 2 - 3 | 312 | 107 | |
GenBank mRNA | BC128872.1 | - | 2 - 3 | 316 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
