Assay Details
Gene Symbol: slc9a6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098256.2 | NP_001091726.2 | 10 - 11 | 1425 | 63 | |
XM_009291141.3 | XP_009289416.1 | 11 - 12 | 1570 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
